Learn More
Bromine liquid, 99.8%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Liquid |
Quantity | 25 g |
Assay Percent Range | 99.8% |
IUPAC Name | bromide |
Formula Weight | 159.81 |
Percent Purity | 99.8% |
Density | 3.1g/cm3 |
Chemical Name or Material | Bromine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T02K019 | Certificate of Analysis | 30/07/2024 | 000905.30,000905.A1,000905.14,000905.D9 |
T07J006 | Certificate of Analysis | 13/07/2023 | 000905.30,000905.A1,000905.14,000905.D9,000905 |
P30I002 | Certificate of Analysis | 12/04/2022 | 000905.30,000905.A1,000905.14,000905.D9 |
Safety and Handling



- Bromine liquid
- Danger
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 2
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P320-Specific treatment is urgent (see on this label).
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.