missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, ACS, 99.5%, Thermo Scientific Chemicals

Product Code. 15461317
Click to view available options
Quantity:
10 g
100 g
500 g
5 x 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 0.005%, Cl 0.05%, I 0.001%, Sulfur compounds (as S) 0.001%, Heavy Metals (as Pb) 2ppm, Ni 5ppm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]

Specifications

Color Brown to Red
Physical Form Liquid
Quantity 100 g
Assay Percent Range 99.5%
IUPAC Name bromide
Formula Weight 159.81
Percent Purity 99.5%
Grade ACS
Odor Unpleasant
Appearance Red-brown fuming liquid
Vapor Pressure 230 hPa (173mm Hg) at 20°C
Density 3.12g/cm3 at 20°C
Chemical Name or Material Bromine liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.