missing translation for 'onlineSavingsMsg'
Learn More

Bromine, puriss. p.a., Honeywell Fluka™

Product Code. 15651530
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
IUPAC Name bromide
SMILES [Br-]

Specifications

Quantity 500 mL
UN Number 1744
Formula Weight 159.81g/mol
Percent Purity ≥99% (RT)
Grade puriss. p.a.
Vapor Pressure 671mmHg (55°C), 175mmHg (20°C)
Density 3.119g/mL at 25°C; Vapor Density: 7.14 (vs air)
Chemical Name or Material Bromine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'