missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, Pure, Indicator Grade, Fisher Chemical™

32.10 € - 3950.00 €

Specifications

Melting Point 225°C
CAS 76-60-8
Molecular Formula C21H14Br4O5S
MDL Number 5874
Formula Weight 698.01g/mol
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10355170
View Documents
Fisher Chemical
B/4320/43
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10223900
View Documents
Fisher Chemical
B/4320/44
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10263180
View Documents
Fisher Chemical
B/4320/46
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

225°C
C21H14Br4O5S
698.01g/mol
Beige
76-60-8
5874
Glass Jar
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bromocresol Green

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.