missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green sodium salt, ACS

37.69 € - 113.57 €

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
MDL Number MFCD00148898
Synonym Bromocresol Green, water soluble
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15476299
View Documents Promotion Details
Thermo Scientific Chemicals
J64284.06
5 g
37.69 €
5g
In Stock
to see stock.
Add to basket
 
15486299
View Documents Promotion Details
Thermo Scientific Chemicals
J64284.14
25 g
113.57 €
25g
In Stock
to see stock.
Add to basket
 
Description

Description

Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

Solubility
40mg/ml in water; 60mg/ml in ethanolSoluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

62625-32-5
C21H13Br4NaO5S
MFCD00148898
Bromocresol Green, water soluble

Specifications

230°C (decomposition)
C21H13Br4NaO5S
14,1386
40mg/ml in water; 60mg/ml in ethanolSoluble in water and ethanol.
ACS
Crystalline Powder
62625-32-5
MFCD00148898
Bromocresol Green, water soluble
720.02
Brown to Red
Bromocresol Green sodium salt
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 263-657-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only