missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple, sodium salt, pure, water soluble, indicator

Product Code. 10791881
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]

Specifications

Beilstein 19, 91
Merck Index 15, 1394
Solubility Information Solubility in water: Soluble
Infrared Spectrum Authentic
Formula Weight 562.2
Grade Pure
Packaging Glass bottle
Specific Extinction 900 min. (590nm, 0.01N NaOH)
Visual Transition Interval from pH 5.2 (yellow) to p 6.8 (purple)
Color Brown to Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromocresol Purple, sodium salt, Water soluble, Indicator, Pure
Show More Show Less
Product Identifier
  • Bromocresol Purple, sodium salt, water soluble, indicator

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'