missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromocresol Purple, sodium salt, pure, water soluble, indicator
27.20 € - 197.00 €
Chemical Identifiers
CAS | 62625-30-3 |
---|---|
Molecular Formula | C21H15Br2NaO5S |
Molecular Weight (g/mol) | 562.204 |
MDL Number | MFCD00148896 |
InChI Key | KVSJYIXUWNOWBD-UGAWPWHASA-M |
Synonym | BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt |
PubChem CID | 23665568 |
IUPAC Name | sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
SMILES | CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10791881
|
Thermo Scientific Chemicals
229610050 |
5 g | Glass bottle |
27.20 €
5g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
10194560
|
Thermo Scientific Chemicals
229610100 |
10 g | Glass bottle |
44.20 €
10g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
10581091
|
Thermo Scientific Chemicals
229610250 |
25 g | Glass bottle |
82.50 €
25g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
10561354
|
Thermo Scientific Chemicals
229611000 |
100 g | Glass bottle |
197.00 €
100g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
62625-30-3 | |
562.204 | |
KVSJYIXUWNOWBD-UGAWPWHASA-M | |
23665568 | |
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+] |
C21H15Br2NaO5S | |
MFCD00148896 | |
BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt | |
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
Specifications
62625-30-3 | |
MFCD00148896 | |
15, 1394 | |
Solubility in water: Soluble | |
Authentic | |
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate | |
23665568 | |
Pure | |
from pH 5.2 (yellow) to p 6.8 (purple) | |
Powder |
C21H15Br2NaO5S | |
19, 91 | |
BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt | |
KVSJYIXUWNOWBD-UGAWPWHASA-M | |
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+] | |
562.204 | |
562.2 | |
900 min. (590nm, 0.01N NaOH) | |
Brown to Red | |
Bromocresol Purple, sodium salt, Water soluble, Indicator, Pure |
RUO – Research Use Only