missing translation for 'onlineSavingsMsg'
Learn More
Bromocycloheptane, 97%, Thermo Scientific Chemicals Product Code.: 11416326

Bromocycloheptane, 97%, Thermo Scientific Chemicals

Product Code. 11416326
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11416326

Brand: Thermo Scientific Alfa Aesar B23110.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2404-35-5
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00004149
InChI Key LOXORFRCPXUORP-UHFFFAOYSA-N
Synonym cycloheptane, bromo, cycloheptyl bromide, cycloheptylbromide, bromocyclo-heptane, 2-bromocycloheptane, bromocycloheptane, acmc-209g8c
PubChem CID 16992
IUPAC Name bromocycloheptane
SMILES BrC1CCCCCC1

Specifications

Density 1.3
Boiling Point 71°C to 72°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.5052
Quantity 25 g
Beilstein 1098389
Formula Weight 177.09
Percent Purity 97%
Chemical Name or Material Bromocycloheptane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10229479 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog NumberB23110.22,B23110.06,B23110,B23110.14
Lot Number10193421 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23110.22,B23110.06,B23110.14
Lot Number10172393 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23110.22,B23110.06,B23110.14
Lot Number10229479 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23110.22,B23110.06,B23110.14
Lot Number10170967 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23110.22,B23110.06,B23110.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromocycloheptane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.