missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 96%, stab. with ethanol, Thermo Scientific Chemicals

Bromoform, 97%, CHBr3, CAS Number-75-25-2, tribromomethane, methenyl tribromide, methane, tribromo, tribrommethan, bromoforme, bromoformio, tribromometan, tribrommethaan, methyl tribromide, rcra waste number u225, 5000g, 1731048, 148 deg.C to 150 deg.C, CHEBI:38682, 2.81

423.00 €

Chemical Identifiers

CAS 75-25-2
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.731
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11403884
View Documents
Thermo Scientific Alfa Aesar
A11904.0I
5000 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.

Solubility
Slightly soluble in water.

Notes
Light-sensitive. A lachrymator. Incompatible with chemically active metals and strong bases.
Specifications

Chemical Identifiers

75-25-2
252.731
DIKBFYAXUHHXCS-UHFFFAOYSA-N
5558
bromoform
CHBr3
MFCD00000128
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
CHEBI:38682
C(Br)(Br)Br

Specifications

75-25-2
2.81
None
1.585
UN2515
Light sensitive
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
DIKBFYAXUHHXCS-UHFFFAOYSA-N
bromoform
5558
252.73
Bromoform, Stabilized with ethanol
8°C to 9°C
148°C to 150°C
CHBr3
MFCD00000128
1731048
14,1420
Slightly soluble in water.
C(Br)(Br)Br
252.731
CHEBI:38682
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302-H332-H315-H319
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c

H302-H315-H319-H331

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BROMOFORM

missing translation for 'einecsNumber' : 200-854-6

missing translation for 'rtecsNumber' : PB5600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.