missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 96%, stabilized, Thermo Scientific Chemicals

77.59 € - 906.51 €

Chemical Identifiers

CAS 75-25-2, 64-17-5
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10256110
View Documents Promotion Details
Thermo Scientific Acros
158202500
250 mL Glass bottle
77.59 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10377570
View Documents Promotion Details
Thermo Scientific Acros
158200010
1 L Glass bottle
220.10 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10573244
View Documents Promotion Details
Thermo Scientific Acros
158200025
2.5 L Plastic drum
486.42 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
13468378
View Documents Promotion Details
Thermo Scientific Acros
158200050
5 L Plastic drum
906.51 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-25-2, 64-17-5
252.73
DIKBFYAXUHHXCS-UHFFFAOYSA-N
5558
bromoform
CHBr3
MFCD00000128
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
CHEBI:38682
C(Br)(Br)Br

Spécification

75-25-2, 64-17-5
Colorless to Orange-Yellow
150.0°C to 151.0°C
95% min. (GC)
CHBr3
01,68
15,143
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
DIKBFYAXUHHXCS-UHFFFAOYSA-N
bromoform
252.73
CHEBI:38682
96%
Bromoform
8.0°C
2.6300g/mL
Authentic
CHBr3
MFCD00000128
12,74
2.63
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene,chloroform,ether,,petroleum ether,acetone and oils
C(Br)(Br)Br
3 to 4 wt% ethanol
5558
252.73
Liquid
Vidéos
Sécurité et traitement

Sécurité et traitement

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-854-6

Mention d'avertissement
  • Danger

Catégorie de danger
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Mention de danger
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Conseils de prudence
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Informations supplémentaires
  • MIXTURE LIST-Contains : Bromoform, Ethyl alcohol

FDS
Documents

Documents

Product Certifications

RUO – Research Use Only