missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoform, 99+%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 8°C |
Color | White |
Density | 2.8900g/mL |
Boiling Point | 150°C to 151°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (excluding stabilizer) (GC) |
Packaging | Glass bottle |
Linear Formula | CHBr3 |
Quantity | 500 mL |
Beilstein | 01, 68 |
Show More |
Safety and Handling
Product Identifier
- Bromoform, stabilized
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H331-Toxic if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : ethanol, bromoform
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction