missing translation for 'onlineSavingsMsg'
Learn More

Bromohydroquinone, 94%, pract., Thermo Scientific™

Product Code. 10239450
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-69-7
MDL Number MFCD00041747
InChI Key REFDOIWRJDGBHY-UHFFFAOYSA-N
Synonym bromohydroquinone, 2-bromohydroquinone, 1,4-benzenediol, 2-bromo, 2-bromoquinol, 2-bromo-1,4-benzenediol, hydroquinone, bromo, bromoquinol, 4-hydroxy bromophenol, 2-bromo-benzene-1,4-diol
PubChem CID 68502
IUPAC Name 2-bromobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Br)O

Specifications

Melting Point 104°C to 110°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C6H5BrO2
Quantity 1 g
Molecular Weight (g/mol) 189.01
Formula Weight 189.01
Percent Purity 94%
Grade Practical
Physical Form Fine Crystalline Powder
Chemical Name or Material Bromohydroquinone
Show More Show Less
compliance-icons
Product Identifier
  • Bromohydroquinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.