missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, Indicator, Pure

Bromophenol Blue, C19H10Br4O5S, CAS Number-115-39-9, 3', 3″, 5', 3',3″,5',5″-Tetrabromophenolsulfonephthalein, BPB, 25g, 19, I, 649, CHEBI:59424, Orange-Pink to Red, 669.96, Pure, UDSAIICHUKSCKT-UHFFFAOYSA-N, Authentic

91.56 € - 261.30 €

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10551591
View Documents
Thermo Scientific Chemicals
151340250
25 g Glass bottle
91.56 €
25g
In Stock
to see stock.
Add to basket
 
10564264
View Documents
Thermo Scientific Chemicals
151341000
100 g Glass bottle
261.30 €
100g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

115-39-9
669.96
UDSAIICHUKSCKT-UHFFFAOYSA-N
8272
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
C19H10Br4O5S
MFCD00005875
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
CHEBI:59424
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

279.0°C
C19H10Br4O5S
19, I, 649
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
UDSAIICHUKSCKT-UHFFFAOYSA-N
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
669.96
CHEBI:59424
Pure
from pH 3.0 (yellow) to pH 4.6 (blue)
Powder
115-39-9
MFCD00005875
15, 1454
Solubility in water: 0.4g/100mL
Authentic
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
8272
669.96
1100 min. (592nm, in buffer)
Orange/Pink to Red
Bromophenol Blue, Indicator
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.