missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, Indicator, Reag. Ph. Eur., Honeywell Fluka™
Indicator, Reag. Ph. Eur.
51.36 € - 172.03 €
Chemical Identifiers
CAS | 115-39-9 |
---|---|
Molecular Formula | C19H10Br4O5S |
Molecular Weight (g/mol) | 669.96 |
MDL Number | MFCD00005875 |
InChI Key | UDSAIICHUKSCKT-UHFFFAOYSA-N |
PubChem CID | 8272 |
ChEBI | CHEBI:59424 |
IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
115-39-9 | |
669.96 | |
UDSAIICHUKSCKT-UHFFFAOYSA-N | |
CHEBI:59424 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br |
C19H10Br4O5S | |
MFCD00005875 | |
8272 | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
Specifications
204°C | |
C19H10Br4O5S | |
NONH for all modes of transport | |
UDSAIICHUKSCKT-UHFFFAOYSA-N | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
8272 | |
669.96g/mol | |
Red | |
Powder |
115-39-9 | |
MFCD00005875 | |
61698 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br | |
669.96 | |
CHEBI:59424 | |
Glass Bottle | |
2.8 to 4.4 | |
Bromophenol Blue, Reag. Ph. Eur. |
Safety and Handling
missing translation for 'einecsNumber' : 204-086-2
missing translation for 'rtecsNumber' : SJ7453000