missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue Sodium Salt Indicator, Honeywell Fluka™
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
UN Number | NONH for all modes of transport |
Beilstein | 6995701 |
Solubility Information | Soluble in water |
Formula Weight | 691.94g/mol |
Packaging | Glass Bottle |
Color | Blue |
pH | 4.6 |
Quantity | 50 g |
Physical Form | Powder |
Chemical Name or Material | Bromophenol Blue Sodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'