missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromophenol Red, pure
71.60 € - 230.00 €
Chemical Identifiers
CAS | 2800-80-8 |
---|---|
Molecular Formula | C19H12Br2O5S |
Molecular Weight (g/mol) | 512.168 |
MDL Number | MFCD00023014 |
InChI Key | OYCLSQDXZMROJK-UHFFFAOYSA-N |
Synonym | 5', 5''-Dibromophenolsulfonphthalein |
PubChem CID | 76047 |
IUPAC Name | 2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10043660
|
Thermo Scientific Chemicals
304550050 |
5 g |
71.60 €
5g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
10359310
|
Thermo Scientific Chemicals
304550250 |
25 g |
230.00 €
25g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2800-80-8 | |
512.168 | |
OYCLSQDXZMROJK-UHFFFAOYSA-N | |
76047 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br |
C19H12Br2O5S | |
MFCD00023014 | |
5', 5''-Dibromophenolsulfonphthalein | |
2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
Specifications
2800-80-8 | |
MFCD00023014 | |
OYCLSQDXZMROJK-UHFFFAOYSA-N | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br | |
512.168 | |
512.19 | |
Pure | |
from pH 5.2 (yellow-red) to pH 7.0 (purple) | |
Powder |
C19H12Br2O5S | |
5', 5''-Dibromophenolsulfonphthalein | |
Authentic | |
2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
76047 | |
5% max. (110°C) | |
Glass bottle | |
Brown to Brown/Red | |
Bromophenol Red |
Safety and Handling
missing translation for 'einecsNumber' : 220-538-1
RUO – Research Use Only