missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromothymol Blue, Pure, Conform to ACS
38.19 € - 306.53 €
Chemical Identifiers
CAS | 76-59-5 |
---|---|
Molecular Formula | C27H28Br2O5S |
Molecular Weight (g/mol) | 624.384 |
MDL Number | MFCD00005872 |
InChI Key | NUHCTOLBWMJMLX-UHFFFAOYSA-N |
Synonym | Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB |
PubChem CID | 6450 |
ChEBI | CHEBI:86155 |
IUPAC Name | 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10209980
|
Thermo Scientific Chemicals
403250050 |
5 g | Glass bottle |
38.19 €
5g |
In Stock
Log in to see stock. |
||||
10219980
|
Thermo Scientific Chemicals
403250100 |
10 g | Glass bottle |
63.32 €
10g |
In Stock
Log in to see stock. |
||||
10422771
|
Thermo Scientific Chemicals
403250250 |
25 g | Glass bottle |
106.53 €
25g |
In Stock
Log in to see stock. |
||||
10658234
|
Thermo Scientific Chemicals
403251000 |
100 g | Glass bottle |
306.53 €
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
76-59-5 | |
624.384 | |
NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
6450 | |
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol |
C27H28Br2O5S | |
MFCD00005872 | |
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB | |
CHEBI:86155 | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br |
Specifications
204.0°C | |
Passes Test | |
MFCD00005872 | |
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB | |
NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br | |
624.384 | |
CHEBI:86155 | |
ACS Reagent | |
Brown/Pink to Purple | |
Bromothymol Blue, Pure |
76-59-5 | |
C27H28Br2O5S | |
15, 1455 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, alkaline solution | |
Authentic | |
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol | |
6450 | |
624.38 | |
from pH 6.0 (yellow) to pH 7.6 (blue) | |
Crystalline Powder |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title