missing translation for 'onlineSavingsMsg'
Learn More

Bromotrichloromethane, 97%

Product Code. 11479772
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11479772

Brand: Thermo Scientific Alfa Aesar A10824.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-62-7 | CBrCl3 | 198.265 g/mol

Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-62-7
Molecular Formula CBrCl3
Molecular Weight (g/mol) 198.265
MDL Number MFCD00000783
InChI Key XNNQFQFUQLJSQT-UHFFFAOYSA-N
Synonym methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br
PubChem CID 6383
IUPAC Name bromo(trichloro)methane
SMILES C(Cl)(Cl)(Cl)Br

Specifications

Melting Point -6°C
Density 2.015
Boiling Point 104°C to 105°C
Flash Point None
Odor Chloroform-like
Linear Formula BrCCl3
Refractive Index 1.506
Quantity 100 g
UN Number UN2810
Beilstein 1732543
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 198.28
Percent Purity 97%
Chemical Name or Material Bromotrichloromethane
Show More Show Less

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado