missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromotrimethylsilane, 97%, stab. with copper powder or silver wire, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.
Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.
Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
Specifications
Specifications
Boiling Point | 78°C to 80°C |
Melting Point | -44°C |
Quantity | 50 g |
Linear Formula | (CH3)3SiBr |
UN Number | UN2924 |
Beilstein | 1731135 |
Flash Point | −3°C (26°F) |
Solubility Information | Miscible with carbon tetrachloride,chloroform,dichloroethane,acetonitrile,toluene,dichloromethane and hexane. |
Formula Weight | 153.09 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'