missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Clonetics™ Bronchial Epithelial Cell Growth Medium Bulletkit

For growth of bronchial/tracheal epithelial cells. Lonza™ Clonetics™ Bronchial Epithelial Cell Growth Medium Bulletkit includes necessary components for primary cell culture.

Brand:  Lonza™ CC-3170


Product Code. 11675440

  • 645.45 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Includes:
  • BEGM™
  • BulletKit™
  • BEBM™
  • plus SingleQuots™ of Growth Supplements
  • 500 ml

Order Info

dimProductType

TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 ml BEBM basal medium plus CC-4175 singlequots kit (formulates 500 ml of BEBM basal medium to BEGM growth medium; contains BPE, 2.0 ml; Hydrocortisone, 0.5mL; hEGF, 0.5mL; Epinephrine, 0.5mL; Transferrin, 0.5mL; Insulin, 0.5mL; Retinoic Acid, 0.5mL; Triiodothyronine, 0.5mL; GA, 0.5mL )to formulate BEGM medium (growth medium)
Growth Medium
1 Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BEGM Bronchial epithelial BulletKit

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol resistant foam to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, gentamicinsulphate

SDS
Documents

Documents

Certificates
Promotions

Promotions