missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brooker's merocyanine dye

Product Code. 15452779
Click to view available options
Quantity:
250 mg
1 g

Brooker's merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brooker′s merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23302-83-2
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00010099
InChI Key DBOHWMPKJCJANT-UHFFFAOYSA-N
Synonym brooker's merocyanine, 4-2-1-methylpyridin-4 1h-ylidene ethylidene cyclohexa-2,5-dien-1-one, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-dien-1-one, 2,5-cyclohexadien-1-one,4-2-1-methyl-4 1h-pyridinylidene ethylidene, brooker/'s merocyanine, 1-methyl-4-4-oxylatostyryl pyridinium, 1-methyl-4-e-4-oxylatostyryl pyridinium, 4-2-1-methylpyridin-4-ylidene ethylidene cyclohexa-2,5-die, 4-2-1-methyl-4-pyridylidene ethylidene cyclohexa-2,5-dien-1-one
PubChem CID 258436
IUPAC Name 4-[2-(1-methylpyridin-4-ylidene)ethylidene]cyclohexa-2,5-dien-1-one
SMILES CN1C=CC(=CC=C2C=CC(=O)C=C2)C=C1

Specifications

Melting Point 220°C (decomposition)
Quantity 1 g
Formula Weight 211.26
Chemical Name or Material Brooker's merocyanine dye

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.