Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2640294
 View more versions of this product
Product Code. 10761041
  • 146.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
Glass
2000 mL
370 mm
29/32 NS Standard Ground
Yes
Brown
Brown
Borosilicate Glass, Duran Glass
±0.6 mL
±0.6 mL
A