Learn More
Hirschmann™ Class A Amber Glass Volumetric Flasks with Poly Stopper
Allows for precise, safe liquid measurement and handling of light-sensitive materials in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks with Poly Stopper are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2640165
 View more versions of this product
Product Code. 11706712
  • 70.75 € / Pack of 2
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
Brown
Borosilicate Glass, Duran Glass
±0.04 mL
±0.04 mL
A
Polyethylene
20 mL
110 mm
10/19 NS Standard Ground
Yes