missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Class A Amber Glass Volumetric Flasks with Poly Stopper
Allows for precise, safe liquid measurement and handling of light-sensitive materials in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks with Poly Stopper are manufactured from durable DURAN™ glass tubing.
69.00 € - 132.00 €
Specifications
Closure Material | Polyethylene |
---|---|
Color | Brown |
Material | Borosilicate Glass, Duran Glass |
Graduated | Yes |
Class | A |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Tolerance | Price | Quantity & Availability | |||
12688665
|
Hirschmann™
2640180 |
100 mL | 12/21 NS Standard Ground | ±0.1 mL |
73.80 €
Pack of 2 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
10598472
|
Hirschmann™
2640193 |
1000 mL | 24/29 NS Standard Ground | ±0.4 mL |
98.90 €
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
11736712
|
Hirschmann™
2640181 |
100 mL | 14/23 NS Standard Ground | ±0.1 mL |
99.10 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
13177939
|
Hirschmann™
2640161 |
10 mL | 10/19 NS Standard Ground | ±0.04 mL |
69.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11796702
|
Hirschmann™
2640160 |
10 mL | 7/16 NS Standard Ground | ±0.025 mL |
77.75 €
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
10761782
|
Hirschmann™
2640194 |
2000 mL | 29/32 NS Standard Ground | ±0.6 mL |
131.00 €
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
10139342
|
Hirschmann™
264016527 |
20 mL | 10/19 NS Standard Ground | ±0.03 mL |
73.30 €
Pack of 2 |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||
11706712
|
Hirschmann™
2640165 |
20 mL | 10/19 NS Standard Ground | ±0.04 mL |
69.00 €
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
11716722
|
Hirschmann™
2640185 |
200 mL | 14/23 NS Standard Ground | ±0.15 mL |
93.10 €
Pack of 2 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||
11726722
|
Hirschmann™
2640186 |
250 mL | 14/23 NS Standard Ground | ±0.15 mL |
105.00 €
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
11652229
|
Hirschmann™
2640170 |
25 mL | 10/19 NS Standard Ground | ±0.04 mL |
69.00 €
Pack of 2 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
10215443
|
Hirschmann™
2640171 |
25 mL | 12/21 NS Standard Ground | ±0.06 mL |
70.20 €
Pack of 2 |
Estimated Shipment: 31-10-2024
Log in to see stock. |
|||
11736722
|
Hirschmann™
2640190 |
500 mL | 19/26 NS Standard Ground | ±0.25 mL |
132.00 €
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
11726712
|
Hirschmann™
2640175 |
50 mL | 12/21 NS Standard Ground | ±0.06 mL |
74.00 €
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
11786702
|
Hirschmann™
2640152 |
5 mL | 7/16 NS Standard Ground | ±0.025 mL |
70.20 €
Pack of 2 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||
11752303
|
Hirschmann™
2640178 |
50 mL | 14/23 NS Standard Ground | ±0.08 mL |
96.60 €
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
11714116
|
Hirschmann™
2640153 |
5 mL | 10/19 NS Standard Ground | ±0.04 mL |
69.00 €
Pack of 2 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
10199192
|
Hirschmann™
264015327 |
5 mL | 10/19 NS Standard Ground | ±0.02 mL |
73.30 €
Pack of 2 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||
Description
- With NS ground and polyethylene stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
Polyethylene | |
Borosilicate Glass, Duran Glass | |
A |
Brown | |
Yes |