Learn More
Brannan™ BS 718 Model Soda Glass M50 SP Petroleum Density Hydrometers
Shot loaded petrolumn density hydrometers for surface tension 0.600 to 0.650g/mL
Brand:  Brannan™ 10/453/0
 View more versions of this product
Product Code. 12831478
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For surface tension 0.600 to 0.650g/mL Shot loaded, length 270mm, subdivisions 0.001g/mL, accuracy ±0.0006g/mL. Calibrated at 15°C.
Specifications
Specifications
1Pack
0.001g/mL
ISO 387, ISO 649
Steel Shot
0.750 to 0.800 g/mL
M50 SP Petroleum Density Hydrometer
0.001 g/mL
270 mm
Petroleum industry and corresponds to IP 160