missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)

Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.

Brand:  Thermo Scientific™ 21580

207.10 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.



 View more versions of this product

Product Code. 10249963

  • 218.00 € / 50mg
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.

Highlights:

  • Crosslink cell-surface proteins before cell lysis and immunoprecipitation
  • Identify receptor-ligand interactions by chemical crosslinking
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Identify near-neighbor protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amine-Amine
572.43
11.4 Å
No
Pierce™
Homobifunctional
Medium (10 to 30 Å)
50 mg
Standard, Single-use, Premium-grade
Crosslinker
No
No
Upon receipt store desiccated at 4°C.
Ambient
Chemical Labeling
Sulfo-NHS Ester
Powder
Water
Yes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PierceBS3,No-Weigh Format

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : BS3

SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.