Learn More
Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent
Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.
Brand: Thermo Scientific™ TS-38831
162.79 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.
- Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
- Catalyzed formulation is stronger than BSTFA alone

Specifications
GC | |
BSTFA +1% TMCS; 10 x 1 mL ampules | |
10 x 1 mL ampules |
98 to 100, 0.1 to 1% | |
2°C to 8°C | |
BSTFA +1% TMCS |
Safety and Handling
- BSTFA + 1% TMCS
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Trimethylsilyl 2, 2,2-trifluoro-N-(trimethylsilyl) acetimidate 25561-30-2 ( 98-100 ), Trimethylchlo
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.