missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA Silylation Reagent

Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.

155.00 € - 426.17 €

Specifications

Standard Concentration 98 to 100%
For Use With GC
Storage 2°C to 8°C
Standard Components Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11557570
View Documents
Thermo Scientific™
TS-38828
25 g
155.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11567570
View Documents
Thermo Scientific™
TS-38829
100 g
426.17 €
Each
Please to purchase this item. Need a web account? Register with us today!
11776957
View Documents
Thermo Scientific™
TS-38830
10 x 1 mL ampules
170.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.

BSTFA is a powerful thrimethylsilyl donor, with donor strength that is comparable to its unfluorinated analog BSA [N,O-Bis(trimethylsilyl)acetamide]. BSTFA reacts to replace labile hydrogens on a wide range of polar compounds with a -Si(CH3)3 group. This physical characteristic is particularly useful in the gas chromatography of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
  • Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA
  • Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA

Specifications

Specifications

98 to 100%
GC
2°C to 8°C
Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BSTFA

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : trimethylsilyl-2,2,2-triflouro-N-(trimethylsilyl) acetimidate

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.