missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BTD Valve Head Complete
Corning™ valve head complete
105.04 € - 138.84 €
Specifications
Description | BTD valve head complete |
---|---|
For Use With (Application) | Dispensing Organic or Inorganic Liquids |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
16802100
|
Corning™
6851 |
2.5 to 10 mL |
105.04 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
16812100
|
Corning™
6852 |
25 to 100 mL |
138.84 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
Description
Specifications
BTD valve head complete | |
Dispensing Organic or Inorganic Liquids |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ BTD Valve Head Complete