missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTEX Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep BTEX
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Benzene | 71-43-2 | 200μg/mL |
Ethylbenzene | 100-41-4 | 200μg/mL |
m-Xylene | 108-38-3 | 200μg/mL |
o-Xylene | 95-47-6 | 200μg/mL |
p-Xylene | 106-42-3 | 200μg/mL |
Toluene | 108-88-3 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
BTEX Standard | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
0.79g/cm3 (at 20°C) | |
Fluid |
2mL amber prescored glass ampule | |
71-43-2, 100-41-4, 108-38-3, 95-47-6, 106-42-3, 67-56-1 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Methanol-P&T |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- BTEX standard
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol
Product intended for laboratory use only