missing translation for 'onlineSavingsMsg'
Learn More

BTS, Tocris Bioscience™

Product Code. 18084172
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of skeletal muscle myosin II ATPase activity

Selective inhibitor of the ATPase activity of skeletal muscle myosin II subfragment 1 (S1) (IC50 ∽ 5 mM). Reversibly blocks gliding motility and suppresses force and twitch production in fast skeletal muscle.

Specifications

CAS 1576-37-0
Molecular Formula C14H15NO2S
Quantity 10 mg
Synonym n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid
InChI Key WTHKAJZQYNKTCJ-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2
IUPAC Name N-benzyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol) 261.339
PubChem CID 95801
Formula Weight 261.34
Percent Purity >99%
Chemical Name or Material BTS
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'