missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes: Handle Only
Handle for use with BTX Two-Needle Array™ Electrodes (cat. no. BTX531).
409.00 € - 447.00 €
Specifications
For Use With (Application) | In-Vivo Drug or Gene Delivery, Muscle Gene Therapy |
---|---|
Material | Delrin |
Product Line | BTX Electroporation |
Product Type | Electroporation accessory |
Voltage | 500 V |
Includes: 8cm long array handle
s
Product Code | Brand | Size (Metric) Gap | Size (English) Gap | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Gap | Size (English) Gap | For Use With (Equipment) | Price | Quantity & Availability | |||
15467290
|
BTX™
450205 |
10 mm | 0.39 in. | 2-needle Array (45-0167), ECM830, Gemini X2 and ECM2001+ Generators |
447.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15477290
|
BTX™
450206 |
5 mm | 0.19 in. | 2-needle Array (45-0168), ECM 830, 2001, 2001+ and Gemini X2 Generators |
409.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Reusable
- 45-0167 handle designed for 45-0120 10 mm needle tips
- Used for larger muscle masses
- 45-0168 handle designed for 45-0121 5 mm needle tips
- Recommended for smaller muscle masses
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
BTX Electroporation | |
500 V | |
1 Ea. | |
Banana Cables |
Delrin | |
Electroporation accessory | |
10 μsec. to 99 msec. in PBS | |
Non-sterile | |
Square Wave |