missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Bubble Sof-Tred™ Dyna-Shield™ Layer with Unique Bubble Design
missing translation for 'orderingAttributeHoverText'
Length (Metric):
91 cm
150 cm
1830 cm
Width (Metric):
60 cm
91 cm
122 cm
missing translation for 'unitSize'
Each
Description
- Lengthwise corrugated rib pattern for sure footing
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Specifications
| Color | Black/Yellow |
| Length (English) | 36 in. |
| Length (Metric) | 91 cm |
| Material | Microcellular Vinyl |
| Type | Industrial and Commercial Matting |
| Width (Metric) | 60 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction