Learn More
BUCHI Boiling Point Tubes and Capillaries
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
Brand:  BUCHI 019697
 View more versions of this product
Product Code. 12725063
  • 53.50 € / Pack of 10
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High precision for reliable resutls
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments
Specifications
Specifications
Boiling Point Tubes
For reproducible boiling point determinations
Borosilicate Glass 3.3
Autoclavable
2.75 mm
Boiling point apparatus
10/Pk.