Learn More
BUCHI Condensers for BUCHI Rotavapors
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
1105.00 € - 2710.00 €
Specifications
For Use With (Application) Evaporator Condensation
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Material (Coating) Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Material (Coating) Type Price Quantity & Availability  
10215222
View Documents Promotion Details
BUCHI
040640
Rotavapor R-210/R-215 None Condenser C
1695.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
12752255
View Documents Promotion Details
BUCHI
040642
Rotavapor R-205, R-210/R-215 Plastic and Glass Condenser C
2710.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
10077051
View Documents Promotion Details
BUCHI
040651
Rotavapor R-210/R-215 None Condenser S
1530.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
10302282
View Documents Promotion Details
BUCHI
040653
Rotavapor R-210/R-215 Plastic and Glass Condenser S
1105.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
Description
Description
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
Specifications
Specifications
Evaporator Condensation