Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044050
 View more versions of this product
Product Code. 12779485
  • 3185.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
70 mm
920 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
218 psi
Column