Learn More
BUCHI Glass Condensers
Sleek cold-trap glass condenser
Brand:  BUCHI 040645
Product Code. 12045502
  • 2685.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Type C condenser is equipped with a cold-trap insert for cooling with dry ice.
Includes:
  • Condenser outer glass
  • Condenser inner
  • Screw caps
  • Hose connections
  • Rubber drip ring
Specifications
Specifications
Type C condenser is equipped with a cold-trap insert for cooling with dry ice
Condenser outer glass, condenser inner, screw caps and hose connections, rubber drip ring
Cold Trap Condenser