missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments

Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.

Brand:  BUCHI 11055014

 View more versions of this product

Product Code. 12033303

  • 111.00 € / Each
Estimated Shipment: 29-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
Specifications

Specifications

Sample Holder
82 x 62 x 30 mm
BUCHI Melting Point Instrument
30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers