Learn More
BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.
Brand:  BUCHI 029508
 View more versions of this product
Product Code. 12747230
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.
  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
Specifications
Specifications
Mini Keyboard
USB PS/2 via adapter
26 mm
282 x 132 x 26 mm
BUCHI Melting Point Instrument