Learn More
Micronova™ BucketBinder™ Multi Bucket System
Micronova™ BucketBinder™ Multi Bucket Systems are a cost efficient, ergonomic multi bucket system that does away with the need for bulky carts and frames.
Brand:  Micronova™ BB-DBL-DBRE
 View more versions of this product
Product Code. 16245152
  • 662.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The BucketBinder™ kit includes 2 or 3 polypropylene buckets with casters and one or two sets of BucketBinders (bucket connectors). The system offers easier cleaning and storage - in both small and large controlled environments and the sterile core.
  • Polyketone ‘hooks’ firmly secure two or three rectangular buckets together allowing scale-up or down as required.
  • Easy clip action allows attachment before or after buckets are filled with disinfectant.
  • Sturdy enough to secure two or three buckets - with up to 15 litres of liquid in each
  • Lightweight – less than 2 lbs per pair – BucketBinders™ hold up to strong disinfectants and can be autoclaved and gamma irradiated
  • Once connected, easily house the W-9 W-92 flat wringers, W-9B BoxWringer™ for string mops or W-94 positive pressure flat mop MegaWringer™ wringer
  • Perfectly complements all the Micronova MZone™ FlatMops™, SnapMops™ and SlimLine™ mops.
  • Bucket liners are easily replaced without completely removing BucketBinders™.
  • BucketBinders™ fit the B-7 AC series rectangular buckets.
  • Autoclavable.
Specifications
Specifications
Mop Bucket
Polypropylene, Polyketone, Stainless Steel
Blue and Red
W-9 Series Wringer
55 cm
Non-sterile
44 L (2 Buckets - 22 L/Ea), 12 gal. (2 Buckets - 6 gal./Ea)
Autoclavable
Cleanroom Surface Cleaning and Disinfecting
37 cm
1 Ea.
65 cm