missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buckets and Lids for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ 4-Place Swinging Bucket Rotor 75-002-000

For use with Sorvall Legend Mach 1.6/1.6R Centrifuges when equipped with 4-Place Swinging Bucket Rotor 75-002-000.

213.06 € - 753.75 €

Specifications

For Use With (Equipment) TTH-400 swing bucket rotor
View More Specs

Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11886510
View Documents
Thermo Scientific™
75002004
Round cap for 400mL bucket, pair
213.06 €
Each
Please to purchase this item. Need a web account? Register with us today!
10317552
View Documents
Thermo Scientific™
75002002
400mL Round Bucket
505.52 €
Each
Please to purchase this item. Need a web account? Register with us today!
10246872
View Documents
Thermo Scientific™
75002001
150mL rectangular bucket
753.75 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For use with Sorvall Legend Mach 1.6/1.6R Centrifuges when equipped with 4-Place Swinging Bucket Rotor 75-002-000.

Choose rectangular or round buckets.

  • Rectangular Bucket (75-002-001) capacity: 150mL
  • Round Bucket (75-002-002) capacity: 400mL
  • Aerosol-tight lids are suitable for biocontainment use

Order Info

Buckets and lids are sold separately. Buckets are sold individually; order correct quantity for your rotor. Lids are sold in pairs.

Specifications

Specifications

TTH-400 swing bucket rotor
Videos
SDS
Documents

Documents

Product Certifications