Learn More
Eppendorf™ Buckets for Rotor S-4x1000
Shop All Eppendorf Products
Click to view available options
Container Type:
High-Capacity Bucket
Plate/Tube Bucket
Round Bucket
Quantity:
Set of 2 pcs.
Set of 4 pcs.
Unit Size:
Pack of 2
Pack of 4
Description
A variety of centrifuge buckets to fit your needs:
- Round bucket S-4x1000 4-piece set
- Round bucket S-4x1000 2-piece set
- Plate/tube bucket S-4x1000 4-piece set
- Plate/tube bucket S-4x1000 2-piece set
- High-capacity bucket S-4x1000 4piece set
- High-capacity bucket S-4x1000 2-pieces
Specifications
Specifications
| For Use With (Equipment) | Rotor S-4x1000 |
| Container Type | High-Capacity Bucket |
| Quantity | Set of 2 pcs. |