Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
1467.30 € - 3648.15 €
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
15600937
|
Thermo Scientific™
75003846 |
12/16 Swinging Bucket Rotors | HAEMAFlex 12/16 swinging bucket rotors |
3648.15 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15880582
|
Thermo Scientific™
75003836 |
Oval, Single Blood Bag Bucket, with Dura-Coat (set of 2) | HAEMAFlex 6/8 swinging bucket rotors |
1547.70 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15729389
|
Thermo Scientific™
75003834 |
Oval, Single, Blood Bag, Bucket, 6/8 Swinging Bucket Rotor | HAEMAFlex 6/8 swinging bucket rotors |
1467.30 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
16 x 500 mL |