Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
972.84 € - 1266.30 €
Specifications
Capacity 4 Standard or 2 Midi-Deepwell plates
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15125283
View Documents
Thermo Scientific™
75005723
Unsealed Bucket
972.84 €
Pack of 2
15241026
View Documents
Thermo Scientific™
75005721
Sealed Bucket
1266.30 €
Pack of 2
Description
Description
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Max Tube Dimensions: Height <33 mm
Specifications
Specifications
4 Standard or 2 Midi-Deepwell plates
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor