missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Bürker Counting Chambers
Shows 9 large squares of 1mm2 each used for counting leucocytes. BRAND™ Bürker Counting Chambers also contain group squares that correspond in size to the Neubauer counting chamber which are used for counting thrombocytes and erythrocytes. CE-marked according to IVD-Directive 98/79 CE.
Brand: Brand™ 718920
This item is not returnable.
View return policy
Description
- Shows 9 large squares of 1mm2 each used for counting leucocytes.
- Each large square is subdivided by double lines (0.05 mm apart) into 16 group squares with 0.2mm sides.
- Group squares correspond in size to the Neubauer counting chamber, but have no further subdivisions.These are used for counting thrombocytes and erythrocytes.
- Double lines from mini squares with an area of 0.0025mm2.
- CE-marked according to IVD-Directive 98/79 CE.
Specifications
Counting Device | |
0.1mm | |
With spring clips |
1Each | |
Counting leucocytes, thrombocytes and erythrocytes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction