missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Concentrated Solution pH8.00 (Borate), NIST Standard for pH measurement, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ea.
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 Ampule |
Packaging | Ampoule |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction