Learn More
IBA Lifesciences Buffer IS for Streptamers
IBA Lifesciences Buffer IS for preparation of wash buffer for the use in Streptamer technology.
Brand:  IBA Lifesciences 6-5602-050
Product Code. 16451485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 10X Concentrated
  • Contains 5 % BSA (w/v) in 10X Phosphate Buffered Saline (PBS)
Specifications
Specifications
50 mL
7.3 to 7.5
50 mL
Wash Buffer
10X
2°C to 8°C
Washing Buffer for Streptamer™ Technology