missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Mercury Free, 25°C, Reagecon™
Click to view available options
Quantity:
1 L
5 L
Color:
Clear
Red
Yellow
pH:
4 ±0.01
7 ±0.01
Description
Used with pH meters to fit pH of the reaction media. Reagecon Buffer, Mercury Free, 25°C are colored buffer solutions with an extended shelf life. Available in pH 4 and 10.
pH values are certified at 25°C to maximize confidence in pH measurements. As well as being ideal for use as calibration standards, they are also cover values that can be used as control standards for all pH applications.
- Mercury free, color-coded pH buffers at 25°C
- Extended shelf life
- Supplied with a detailed certificate of analysis, which outlines traceability to NIST
- Temperature dependence data is printed on label, as are lot numbers and expiry dates
Specifications
Specifications
Color | Clear |
Certificate of Analysis | Supplied |
pH | 4 ±0.01 |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | Bottle |
Traceability to NIST | Traceable to NIST |
Solubility Information | Fully miscible |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'