missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey Nagel Bioanalysis™ Buffer RL1 Macherey-Nagel
Brand: Macherey Nagel Bioanalysis™ 740385.125
This item is not returnable.
View return policy
Specifications
125 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Buffer RL1 (125 mL)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidium thiocyanide