missing translation for 'onlineSavingsMsg'
Learn More
compliance-icons

Technical Buffer pH 9.21 Sachets, Mettler Toledo™
GREENER_CHOICE

Product Code. 12137800 Shop All Mettler Toledo Products
30 x 20 mL
Click to view available options
Quantity:
30 x 20 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Traceable to the primary standards of reference materials selected by NIST.

  • Sachets guarantee fresh solution for every calibration
  • Easy to use
Greener Choice Claims
  • Environmental benefits include:
    • Certifications and Seals of Approval
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

pH 9.21
Quantity 30 x 20 mL
Packaging 30Pack
Traceability to NIST Yes
compliance-icons
Product Identifier
  • Buffer solution pH 9.21
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : disodium tetraboratedecahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Technical Buffer pH 9.21 Sachets, Mettler Toledo™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.